missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HB-EGF Mouse anti-Human, Clone: 406316, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2591-100

This item is not returnable.
View return policy
Description
HB-EGF Monoclonal specifically detects HB-EGF in Human samples. It is validated for Western Blot, Neutralization.Specifications
HB-EGF | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), Dtr, DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), Dts, DTSF, HBEGF, Hegfl, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1839 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Neutralization | |
406316 | |
Western Blot 1 ug/mL, Neutralization 0.5-2.5 ug/mL | |
Q53H93 | |
HBEGF | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HB-EGF Asp63-Leu148 Accession # Q53H93 | |
100 μg | |
Primary | |
Detects human HB-EGF in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Amphiregulin, rhBTC, rhHRG-alpha , rhHRG-beta , rhEGF, rhTGF-alpha , recombinant mouse (rm) Epigen, or rmEpiregulin. | |
Human | |
Supernatant |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
HB-EGF Mouse anti-Human, Clone: 406316, R&D Systems™
Spot an opportunity for improvement?Share a Content Correction