missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A1, Mouse, Clone: BC1, Abnova™
Mouse monoclonal antibody raised against MS4A1.
Brand: Abnova MAB4973.100ug
This item is not returnable.
View return policy
Description
This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. This gene encodes a B-lymphocyte surface molecule which plays a role in the development and differentiation of B-cells into plasma cells. This family member is localized to 11q12, among a cluster of family members. Alternative splicing of this gene results in two transcript variants which encode the same protein. [provided by RefSeq
Specifications
MS4A1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
MS4A1 | |
MS4A1 | |
Normal human lymphocytes from lymph node. | |
RUO | |
931 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgM |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
BC1 | |
Mouse monoclonal antibody raised against MS4A1. | |
In buffer containing 1% BSA, pH 7.2 (0.09% sodium azide) | |
B1/Bp35/CD20/LEU-16/MGC3969/MS4A2/S7 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Liquid |