missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgG2a Secondary Antibody, FITC, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen SA125265
This item is not returnable.
View return policy
Description
SA1-25265 detects rat IgG2a, no cross reactivity is observed with other rat immunoglobulin subclasses. Use 10 μL of the suggested dilution to label 10^6 cells in 100 μL. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat IgG2a | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide | |
Rat IgG2a | |
125 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG1 |
Flow Cytometry | |
MARG2a-1 | |
FITC | |
Mouse | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction