missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse BD-2 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
818.00 SEK - 30750.00 SEK
Specifications
Accession Number | P82020 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 13215 |
Molecular Weight (g/mol) | 5.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17831893
|
Gibco™
250-40-100UG |
100 μg |
6150.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17811003
|
Gibco™
250-40-1MG |
1 mg |
30750.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870623
|
Gibco™
250-40-20UG |
20 μg |
2015.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17871693
|
Gibco™
250-40-250UG |
250 μg |
10770.00 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17860063
|
Gibco™
250-40-500UG |
500 μg |
18430.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17861093
|
Gibco™
250-40-5UG |
5 μg |
818.00 SEK
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.Specifications
P82020 | |
protein with no preservative | |
5.5 | |
E. coli | |
RUO | |
BD-2; beta defensin 2; beta defensin-2; Beta-defensin 2; Beta-defensin 4 A; DEFB102; DEFB2; DEFB-2; DEFB4; DEFB4A; DEFB4B; Defensin beta 2; defensin beta 4 A; Defensin beta2; defensin, beta 2; defensin, beta 4; defensin, beta 4 A; Defensin-beta2; HBD-2; SAP1; Skin-antimicrobial peptide 1 | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
13215 | |
Mouse BD-2 | |
-20°C | |
<1 EU/μg | |
Defb2 | |
Unconjugated | |
AVGSLKSIGY EAELDHCHTN GGYCVRAICP PSARRPGSCF PEKNPCCKYM K | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |