missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse BD-3, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
2320.00 SEK - 33780.00 SEK
Specifications
For Use With (Application) | Functional Assay |
---|---|
Formulation | protein with no preservative |
Molecular Weight (g/mol) | 4.6 |
Name | Mouse BD-3, Animal-Free |
Source | E. coli |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17803233
|
Gibco™
AF-250-41-100UG |
100 μg |
7000.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17843623
|
Gibco™
AF-250-41-1MG |
1 mg |
33780.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17863063
|
Gibco™
AF-250-41-20UG |
20 μg |
2320.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17853073
|
Gibco™
AF-250-41-500UG |
500 μg |
20250.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].Specifications
Functional Assay | |
4.6 | |
E. coli | |
RUO | |
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103 A; defensin, beta 103; defensin, beta 103 A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
Unconjugated | |
KKINNPVSCL RKGGRCWNRC IGNTRQIGSC GVPFLKCCKR K | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
protein with no preservative | |
Mouse BD-3, Animal-Free | |
-20°C | |
<0.1 EU/μg | |
Mouse | |
Recombinant | |
Lyophilized |