Learn More
R&D Systems Mouse Luminex™ Discovery Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems LXSAMSM-10
 View more versions of this product
Product Code. 18732134
  • 21365.00 SEK / 1 set
Explore more special offers
This item is not returnable. View return policy
Specifications
10 Plex
Mouse
Multiplex Cytokine Assay Kits
Store the unopened product at 2 - 8 °C. Do not use past expiration date.