Learn More
Mouse T Cell Inhibitory Receptor (Blocking) Antibody Pack, Novus Biologicals™
nan Monoclonal Antibody
Brand: Novus Biologicals NBP3-12531
This item is not returnable.
View return policy
Description
Mouse T Cell Inhibitory Receptor (Blocking) Monoclonal antibody specifically detects Mouse T Cell Inhibitory Receptor (Blocking) in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF, NeutralizationSpecifications
Mouse T Cell Inhibitory Receptor (Blocking) | |
Monoclonal | |
See individual datasheets | |
See individual datasheets | |
RUO | |
Storage is content dependent. |
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF, Neutralization | |
Unconjugated | |
CTLA-4 blocking, LAG-3 blocking, PD-L1 blocking, T cell inhibitor blocking, T cell inhibitor neutralization, TIGIT blocking | |
4 Vials | |
Mouse | |
Purified |