missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CSM-LEU-TRP-URA, Powdered Media
Dropout supplement for S. cereviseae
Brand: MP Biomedicals™ 114530912
This item is not returnable.
View return policy
Description
- Triple dropout
- Complete supplement mixture of amino acids
- Complete formulation
- Dropout formulation
Yeast cell culture, Gene expression, Transcription regulation

Specifications
Molecular biology Certified™ | |
Powder | |
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tyrosine 50mg/L, L-Valine 140mg/L | |
10 g |
15°C to 30°C | |
Yeast Media | |
For growth of Saccharomyces cerevisiae |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ CSM-LEU-TRP-URA, Powdered Media > 10g
Spot an opportunity for improvement?Share a Content Correction