missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ γ-Globulins, Labile Enzyme Free, 10g
Brand: MP Biomedicals™ 08820423
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
10 g | |
Immunoassay | |
Labile Enzyme-free gamma Globulin |
0°C to 5°C | |
>98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction