Learn More
MP Biomedicals™ Lotus tetragonolobus Agglutinin
Exhibits a specificity for α-L-fucose.
Brand:  MP Biomedicals™ 0215330305
Product Code. 11438701
  • 1100.00 SEK / 5mg
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 mg
Powder
L-fucose
-20°C
Lotus tetragonolobus