missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Whole Serum, Hamster, Purified, 2mL

Whole Hamster Serum with buffer salts

Brand:  MP Biomedicals™ 0855986

Product Code. 17367459

  • 1118.30 SEK / 2mL
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C
Hamster Serum
0.2 μm filtration
Yes
62.1mg/mL
Hamster
2 mL
Serum Protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques, used as a blocking agent in fluorescence and enzyme immunoassays
7.3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers