missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Whole Serum, Monkey, Purified, 2mL

Whole Monkey Serum with buffer salts

Brand:  MP Biomedicals™ 0855988

Product Code. 11454315

  • 1060.00 SEK / 2mL
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C
Monkey Serum
Serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques
63.9mg/mL
Monkey
2 mL
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers