missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Yeast Lytic Enzyme

From Arthrobacter luteus

Brand:  MP Biomedicals™ 0219012310

Product Code. 11404572

  • 6530.00 SEK / 10ku

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Activity: 100,000U/g. Unit Definition: One unit will produce a Δ A800 of 0.001 per minute at pH 7.5 and 25°C, using a suspension of brewers yeast as a substrate in a 3mL reaction mixture. This enzyme catalyzes the lysis of yeast cells and hydrolyzes glucose polymers with β-1,3-linkages (e.g., glucan). It is also useful for preparing yeast spheroplasts. Lyophilized powder.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

≥100,000 U/g
5 to 10
0°C
10 U
Lyophilized Powder
Beige
Cell lysis, spheroplasting and Glucan hydrolysis
Arthrobacter luteus
Yeast Lytic Enzyme
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers