Learn More
MRPL38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15913-1-AP-150UL
 View more versions of this product
Product Code. 16825314
  • 4652.90 SEK / 150µL
Explore more special offers
Description
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
Specifications
MRPL38
Polyclonal
Unconjugated
MRPL38
HSPC262, KIAA1863, L38mt, MRP L3, MRP L38, MRPL38, RPML3
Rabbit
Antigen Affinity Chromatography
RUO
303685, 60441, 64978
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5PQN9, Q8K2M0, Q96DV4
Mrpl38
MRPL38 Fusion Protein Ag8706
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Title
MRPL38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated