missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mTOR Monoclonal Antibody (215Q18), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen AHO1232
This item is not returnable.
View return policy
Description
FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.Specifications
mTOR | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
P42345, P42346, Q9JLN9 | |
MTOR | |
Recombinant fragment of human mTOR expressed in E. coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
215Q18 | |
Unconjugated | |
MTOR | |
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS | |
Mouse | |
Affinity chromatography | |
RUO | |
2475, 56717, 56718 | |
-20°C | |
Liquid |