missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories
Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Brand: Thermo Scientific™ 24073002
This item is not returnable.
View return policy
Description
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories.
Notes:
- Cat. No. N15133 to N15139 are to be used with the Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
- Cat. No. 24073292, 24073001, 24073297, 24073002, 24072675, 24072678 and 24072676 are to be used with the Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM
Specifications
Tube Dispensing Cassette | |
SMART Small tube metal tip dispensing cassette 5-pack | |
0.5 to 50 μL | |
<1 mL |
Autoclavable (10 times at 121°C, 1 bar, 20 min.) | |
Stainless Steel Tip, Silicone Tubing | |
Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM | |
Tubing: 0.4 mm; Tip 0.22 mm |