missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories
Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Brand: Thermo Scientific™ N15134
This item is not returnable.
View return policy
Description
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories.
Designed for use with Multidrop Combi SMART+ dispensers Equipped with radio frequency identification (RFID) tags to store lifetime dispensing data and display remaining usage time Available in standard and small tube versions Distinguished from normal cassettes by antenna tags
- Cat. No. N15133 to N15139 are to be used with the Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
- Cat. No. 24073292, 24073001, 24073297, 24073002, 24072675, 24072678 and 24072676 are to be used with the Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM
Specifications
Tube Dispensing Cassette | |
SMART 2 Small tube plastic tip dispensing cassette, new RFID tag, 5 pack | |
0.5 to 50 μL | |
<1 mL |
Autoclavable (10 times at 121°C, 1 bar, 20 min.) | |
Polypropylene Tip, Silicone Tubing | |
Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards | |
Tubing: 0.4 mm; Tip 0.22 mm |