Learn More
Hanna Instruments™ Multiparameter Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Multiparameter Chemical Test Kit features pre-made, pre-measured titration reagents for the determination of multiple parameters commonly measured for environmental/facility monitoring.
1760.00 SEK - 8250.00 SEK
Product Code | Brand | For Use With (Application) | No. of Tests | Sufficient For | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | No. of Tests | Sufficient For | Price | Quantity & Availability | |||
11672360
|
Hanna Instruments™
HI3817 |
Water Quality Monitoring | 5 parameters x 100 tests, 1 parameter (iron) x 50 tests | Testing of: alkalinity, chloride, hardness, iron, pH, and sulfite |
6920.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11666790
|
Hanna Instruments™
HI3896 |
NPK Soil Testing | 4 parameters x 25 tests | Testing of: pH, nitrogen (N), phosphorus (P), and potassium (K) |
1760.00 SEK
Pack of 100 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||
11632370
|
Hanna Instruments™
HI3821 |
Cooling and Boiler Monitoring | 5 parameters x 100 tests each, 1 parameter (phosphate) x 50 tests | Testing of: alkalinity, chloride, hardness, dissolved oxygen, phosphate, and sulfite |
8250.00 SEK
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||
11662370
|
Hanna Instruments™
HI3823 |
Marine / Aquaculture Monitoring | 6 parameters x 100 tests each | Testing of: alkalinity, carbon dioxide, hardness, dissolved oxygen, pH, and salinity |
6820.00 SEK
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||
11632360
|
Hanna Instruments™
HI3814 |
Environmental Monitoring | 6 parameters x 100 tests each | Testing of: pH, acidity, alkalinity, carbon dioxide, hardness, dissolved oxygen |
6320.00 SEK
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, a case and pre-measured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
1 Kit | |
Various |
Safety and Handling
- water test kit
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Flammable liquid Category 2
- Acute toxicity Category 2
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Germ cell mutagenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin sensitiser Category 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H331-Toxic if inhaled.
- H350-May cause cancer.
- H225-Highly flammable liquid and vapour.
- H300-Fatal if swallowed.
- H290-May be corrosive to metals.
- H373-May cause damage to organs.
- H372-Causes damage to organs.,Causes damage to organs.
- H302-Harmful if swallowed.,Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.,Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.,Suspected of causing genetic defects.
- H319-Causes serious eye irritation.,Causes serious eye irritation.
- H311-Toxic in contact with skin.,Toxic in contact with skin.
- H318-Causes serious eye damage.,Causes serious eye damage.
- H361f-Suspected of damaging fertility.,Suspected of damaging fertility.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.,Suspected of damaging fertility. Suspected of damaging the unborn child.
- H332-Harmful if inhaled.,Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.,Harmful to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.,May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.,Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.,Toxic to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
- EUH031-Contact with acids liberates toxic gas.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
- P391-Collect spillage.,Collect spillage.
- P243-Take precautionary measures against static discharge.,Take precautionary measures against static discharge.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.,IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.,IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.,Call a POISON CENTER/doctor/if you feel unwell.
- P370+P378-In case of fire: Use powder to extinguish.,In case of fire: Use powder to extinguish.
- P310-Immediately call a POISON CENTER/doctor/.,Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.,IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P235-Store in a well-ventilated place. Keep cool.,Store in a well-ventilated place. Keep cool.
- P390-Absorb spillage to prevent material damage.,Absorb spillage to prevent material damage.
- P308+P313-IF exposed or concerned: Get medical advice/attention.,IF exposed or concerned: Get medical advice/attention.
- P404-Store in a closed container.,Store in a closed container.
- P337+P313-If eye irritation persists: Get medical advice/attention.,If eye irritation persists: Get medical advice/attention.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : ethanol, phenolphtalein, potassium disulphate, sulfanilic acid, cadmium, 2,5-dihydroxybenzoic acid,
Your input is important to us. Please complete this form to provide feedback related to the content on this product.