Learn More
Thermo Scientific™ Chemiluminescence Exposure Screen for myECL™ Imager
For use with Thermo Scientific myECL Imager
Brand: Thermo Scientific™ 62242
Additional Details : Weight : 0.50000kg
Description
Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.
Replacement parts for the myECL Imager and accessories to expand upon its capabilities.
Specifications
24.5 cm x 19.5 cm x 3.2 mm | |
Transilluminator Screen | |
Chemiluminescence Exposure Screen |
myECL Imager | |
myECL |