missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase Monoclonal Antibody (SP72), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA516383
This item is not returnable.
View return policy
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.Specifications
Myeloperoxidase | |
Monoclonal | |
0.212 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
P05164 | |
MPO | |
Synthetic peptide corresponding to C-terminus of human myeloperoxidase protein. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
SP72 | |
Unconjugated | |
MPO | |
84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
Rabbit | |
Protein A | |
RUO | |
4353 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction