Learn More
Myosin heavy chain 11 Antibody (160), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Bio-Techne NBP2-89243V
Product Code. 18323381
  • 6010.00 SEK / 0.10mL
Explore more special offers
Description
Description
Myosin heavy chain 11 Monoclonal specifically detects Myosin heavy chain 11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Myosin heavy chain 11
Monoclonal
DyLight 405
AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Myosin heavy chain 11.
0.1 mL
Hypoxia, Stem Cell Markers
4629
Store at 4°C in the dark.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
160
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified