missing translation for 'onlineSavingsMsg'
Learn More

N,1-Di-Fmoc-L-histidine, 95%, Thermo Scientific™

Product Code. 15489368
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98929-98-7
Molecular Formula C36H29N3O6
Molecular Weight (g/mol) 599.64
MDL Number MFCD00070658
InChI Key WTBXFPFCUZKREB-UHFFFAOYNA-N
Synonym fmoc-his fmoc-oh, n,n'-bis 9-fluorenylmethyloxycarbonyl-l-histidine, s-3-1-9h-fluoren-9-yl methoxy carbonyl-1h-imidazol-4-yl-2-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, l-histidine, n,1-bis 9h-fluoren-9-ylmethoxy carbonyl, fmoc-his fmoc, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-9h-fluoren-9-ylmethoxy carbonyl imidazol-4-yl propanoic acid, ambotzfaa1497, fmoc-l-his fmoc-oh, pubchem10020, fmoc-histidine fmoc-oh
PubChem CID 45072174
SMILES OC(=O)C(CC1=CN(C=N1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
IUPAC Name 3-(1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-1H-imidazol-4-yl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Formula Weight 599.63
Percent Purity 95%
Chemical Name or Material N,1-Di-Fmoc-L-histidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.