Learn More
N-Acetyl-5-hydroxytryptamine, Thermo Scientific™
Brand:  Thermo Scientific Acros 226930010
Product Code. 17720629
  • 4334.01 SEK / 1g
Explore more special offers
Specifications
N-Acetyl-5-hydroxytryptamine
N-Acetylserotonin
119°C to 123°C
Life science
Powder
≥97.5%
1210-83-9
1 g
No special requirements
C12 H14 N2 O2
White to Light-yellow