missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-Calicheamicin, MedChemExpress
MedChemExpress N-Acetyl-Calicheamicin is a potent enediyne antitumor antibiotic.
Brand: Medchem Express HY-19791

This item is not returnable.
View return policy
Description
N-Acetyl-Calicheamicin is a potent enediyne antitumor antibiotic.Specifications
N-Acetyl-Calicheamicin | |
108212-76-6 | |
1 mg | |
C57H76IN3O22S4 | |
N-Acetyl-Calicheamicin γ N-Acetyl-γ-calicheamicin | |
O[C@](CC1=O)(C#C/C=C\C#C2)/C(C([C@H]2O[C@@](O[C@H](C)[C@@H](NO[C@H](O[C@H](C)[C@H]3SC(C(C(C)=C(I)C(O[C@H](O[C@@H](C)[C@H](O)[C@H]4OC)[C@@H]4O)=C5OC)=C5OC)=O)C[C@@H]3O)[C@@H]6O)([H])[C@@H]6O[C@@](OC[C@@H]7N(CC)C(C)=O)([H])C[C@@H]7OC)=C1NC(OC)=O)=C/CSSSC | |
1410.39 | |
Research |
Solid | |
White | |
Research | |
Cancer-programmed cell death | |
DMSO : 100 mg/mL (70.90 mM; Need ultrasonic) | |
1410.39 | |
98.0% |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
N-Acetyl-Calicheamicin, MedChemExpress
Research purposes only
Spot an opportunity for improvement?Share a Content Correction