missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-Galactosamine, MP Biomedicals™

Product Code. 11328349
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

A component of many O-linked and N-linked glycan structures. N-Acetyl-D-Galactosamine, MP Biomedicals is an aminosugar that is the initial O-linked sugar to many serine and threonine residues in protein glycosylations.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14215-68-0
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.209
InChI Key OVRNDRQMDRJTHS-JAJWTYFOSA-N
Synonym n-acetyl-d-galactosamine, galnac, 2-acetamido-2-deoxy-d-galactopyranose, d-galnac, 2-acetamido-2-deoxy-d-galactose, n-acetyl-d-chondrosamine, n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide, n-acetylgalactosamine, acetylgalactosamine, n-acetyl-galactosamine
PubChem CID 440552
ChEBI CHEBI:28497
IUPAC Name N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)O

Specifications

Melting Point 160°C to 161°C (literature)
Quantity 100 mg
Solubility Information Soluble in water (100mg/mL - clear, colorless solution).
Optical Rotation +84–89° (c=1, water)
Formula Weight 221.21
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.