Learn More
N(alpha)-Benzyloxycarbonyl-D-tryptophan, 98+%, Thermo Scientific™
Product Code. 11392696
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2279-15-4
Molecular Formula C19H18N2O4
Molecular Weight (g/mol) 338.36
MDL Number MFCD00037945
InChI Key AHYFYYVVAXRMKB-QGZVFWFLSA-N
Synonym z-d-trp-oh, n-cbz-d-tryptophan, z-d-tryptophan, cbz-d-trp-oh, nalpha-cbz-d-tryptophan, r-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoic acid, n a-benzyloxycarbonyl-d-tryptophan, chembl65134, nalpha-carbobenzoxy-d-tryptophan, d-tryptophan, n-phenylmethoxy carbonyl
PubChem CID 700663
IUPAC Name (2R)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OC(=O)[C@@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1
Specifications
Melting Point 122°C to 124°C
Beilstein 96744
Quantity 1 g
Formula Weight 338.37
Optical Rotation −3.1° (c=3 in Acetic Acid)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-D-tryptophan