missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-valine, 99%, Thermo Scientific Chemicals

Product Code. 11490104
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1149-26-4
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00008922
InChI Key CANZBRDGRHNSGZ-NSHDSACASA-N
Synonym n-carbobenzyloxy-l-valine, n-cbz-l-valine, z-val-oh, cbz-l-valine, n-benzyloxycarbonyl-l-valine, n-carbobenzoxy-l-valine, z-l-val-oh, carbobenzoxy-l-valine, z-valine, carbobenzyloxy-l-valine
PubChem CID 726987
IUPAC Name (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 60°C
Quantity 100 g
Beilstein 2056617
Formula Weight 251.28
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-valine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'