missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,3-diaminopropane hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11348836
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127346-48-9
Molecular Formula C8H19ClN2O2
Molecular Weight (g/mol) 210.702
MDL Number MFCD01076598
InChI Key WUXOJNUZYOFBMI-UHFFFAOYSA-N
Synonym tert-butyl 3-aminopropyl carbamate hydrochloride, n-boc-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid tert-butyl ester hydrochloride, n-boc-1,3-propanediamine hydrochloride, tert-butyl n-3-aminopropyl carbamate hydrochloride, acmc-20ak6b, n-boc-1,3-diaminopropane hcl, n-3-aminopropyl carbamic acid tert-butyl ester-hcl, tert-butyl 3-aminopropyl carbamatehydrochloride, n-3-aminopropyl carbamic acid tert-butyl ester hcl
PubChem CID 21982912
IUPAC Name tert-butyl N-(3-aminopropyl)carbamate;hydrochloride
SMILES CC(C)(C)OC(=O)NCCCN.Cl

Specifications

Melting Point 159°C to 161°C
Beilstein 3627894
Formula Weight 210.71
Percent Purity 98%
Quantity 250 mg
Sensitivity Hygroscopic
Chemical Name or Material N-Boc-1,3-diaminopropane hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'