missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-1,3-propanediamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10483083
Click to view available options
1g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 127346-48-9
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (Argentometry)
Molecular Formula C8H18N2O2·HCl
Linear Formula (CH3)3CHOC(O)NH(CH2)3NH2·HCl
MDL Number MFCD01076598
Synonym tert-butyl 3-aminopropyl carbamate hydrochloride, n-boc-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid tert-butyl ester hydrochloride, n-boc-1,3-propanediamine hydrochloride, tert-butyl n-3-aminopropyl carbamate hydrochloride, acmc-20ak6b, n-boc-1,3-diaminopropane hcl, n-3-aminopropyl carbamic acid tert-butyl ester-hcl, tert-butyl 3-aminopropyl carbamatehydrochloride, n-3-aminopropyl carbamic acid tert-butyl ester hcl
InChI Key WUXOJNUZYOFBMI-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NCCCN.Cl
IUPAC Name tert-butyl N-(3-aminopropyl)carbamate;hydrochloride
Molecular Weight (g/mol) 210.71
PubChem CID 21982912
Formula Weight 210.71
Percent Purity 97%
Chemical Name or Material N-BOC-1, 3-propanediamine hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'