missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-methyl-L-phenylalanine, 98%, Thermo Scientific Chemicals

1620.00 SEK - 5450.00 SEK

Chemical Identifiers

CAS 114873-06-2
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.336
MDL Number MFCD01862944
InChI Key HBBXWMALJNZDOM-LBPRGKRZSA-N
Synonym boc-phe 3-me-oh, boc-3-methyl-l-phenylalanine, boc-l-3-methylphenylalanine, boc-l-3-methylphe, boc-3-methy-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-l-3-methyl phenylalanine, boc-phe 3-me-oh tlc, 3-methyl-n-t-butoxycarbonyl-l-phenylalanineShow MoreShow Less
PubChem CID 2761470
IUPAC Name (2S)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15409318
SDS View Documents
Thermo Scientific Alfa Aesar
H62860.03
1 g N/A N/A N/A
15419318
SDS View Documents
Thermo Scientific Alfa Aesar
H62860.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-3-methyl-L-phenylalanine, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.