missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl methyl sulfide, 98+%, Thermo Scientific™

Product Code. 11398575
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-29-5
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00015256
InChI Key WCXXISMIJBRDQK-UHFFFAOYSA-N
Synonym butyl methyl sulfide, butane, 1-methylthio, n-butyl methyl sulfide, butyl methyl sulphide, sulfide, butyl methyl, 1-methylthio butane, unii-2n5imb9xap, methyl butyl sulfide, 2-thiahexane, 2n5imb9xap
PubChem CID 12339
IUPAC Name 1-methylsulfanylbutane
SMILES CCCCSC

Specifications

Density 0.85
Boiling Point 122°C to 123°C
Flash Point 20°C (68°F)
Odor Unpleasant
Linear Formula CH3S(CH2)3CH3
Refractive Index 1.4621
Quantity 5 g
UN Number UN1993
Beilstein 1730959
Formula Weight 104.22
Percent Purity ≥98%
Chemical Name or Material n-Butyl methyl sulfide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'