missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ n-Dodecanethiol gold nanoparticles, 2% (w/v) soln. in toluene, 2-5nm particles
Click to view available options
Quantity:
5 mL
Specifications
Specifications
CAS | 13149-93-4 |
MDL Number | MFCD09953506 |
Quantity | 5 mL |
UN Number | UN1294 |
Synonym | Gold-n-dodecanethiol in toluene |
Concentration or Composition (by Analyte or Components) | 2% (w/v) soln. in toluene |
Chemical Name or Material | n-Dodecanethiol gold nanoparticles, 2 to 5nm particles |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'