missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Dodecyl methyl sulfide, Thermo Scientific™
449.00 SEK - 1720.00 SEK
Chemical Identifiers
CAS | 3698-89-3 |
---|---|
Molecular Formula | C13H28S |
Molecular Weight (g/mol) | 216.43 |
MDL Number | MFCD00015073 |
InChI Key | KJWHJDGMOQJLGF-UHFFFAOYSA-N |
Synonym | dodecyl methyl sulfide, 2-thiatetradecane, 1-methylsulfanyl dodecane, methylthiododecane, methyl lauryl sulfide, dodecane, 1-methylthio, methyl n-dodecyl sulfide, dodecyl methyl sulphide, sulfide, dodecyl methyl, unii-77bo1dv08u |
PubChem CID | 77287 |
IUPAC Name | 1-methylsulfanyldodecane |
SMILES | CCCCCCCCCCCCSC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3698-89-3 | |
216.43 | |
KJWHJDGMOQJLGF-UHFFFAOYSA-N | |
77287 | |
CCCCCCCCCCCCSC |
C13H28S | |
MFCD00015073 | |
dodecyl methyl sulfide, 2-thiatetradecane, 1-methylsulfanyl dodecane, methylthiododecane, methyl lauryl sulfide, dodecane, 1-methylthio, methyl n-dodecyl sulfide, dodecyl methyl sulphide, sulfide, dodecyl methyl, unii-77bo1dv08u | |
1-methylsulfanyldodecane |
Specifications
3698-89-3 | |
Sulphurous | |
CH3(CH2)11SCH3 | |
MFCD00015073 | |
KJWHJDGMOQJLGF-UHFFFAOYSA-N | |
1-methylsulfanyldodecane | |
77287 | |
Liquid |
163°C to 165°C (19 mmHg) | |
C13H28S | |
1.4608 | |
dodecyl methyl sulfide, 2-thiatetradecane, 1-methylsulfanyl dodecane, methylthiododecane, methyl lauryl sulfide, dodecane, 1-methylthio, methyl n-dodecyl sulfide, dodecyl methyl sulphide, sulfide, dodecyl methyl, unii-77bo1dv08u | |
CCCCCCCCCCCCSC | |
216.43 | |
216.41 | |
n-Dodecyl methyl sulfide |
Safety and Handling
missing translation for 'einecsNumber' : 223-028-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title