missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecylpyridinium chloride hydrate, 98%, Thermo Scientific Chemicals

Product Code. 11445497
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207234-02-4
Molecular Formula C17H30ClN
Molecular Weight (g/mol) 283.88
MDL Number MFCD00011987
InChI Key GKQHIYSTBXDYNQ-UHFFFAOYSA-M
Synonym 1-dodecylpyridin-1-ium chloride hydrate, laurylpyridinum chloride, 1-dodecylpyridinium chloride hydrate, 1-dodecylpyridin-1-ium hydrate chloride, c17h30n.cl.h2o, n-dodecylpyridinium chloride hydrate, dodecylpyridinium chloride monohydrate, n-dodecylpyridinium chloride monohydrate, 1-dodecylpyridin-1-ium chloride-water 1/1/1
PubChem CID 16211802
IUPAC Name 1-dodecylpyridin-1-ium;chloride;hydrate
SMILES [Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1

Specifications

Melting Point 88°C to 90°C
Quantity 100 g
Sensitivity Hygroscopic
Formula Weight 283.89 (Anhydrous)
Percent Purity 98%
Chemical Name or Material n-Dodecylpyridinium chloride hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'