missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-L-alanine, 98%, Thermo Scientific Chemicals
295.00 SEK - 795.00 SEK
Chemical Identifiers
CAS | 3913-67-5 |
---|---|
Molecular Formula | C4H9NO2 |
Molecular Weight (g/mol) | 103.121 |
MDL Number | MFCD00037241 |
InChI Key | GDFAOVXKHJXLEI-VKHMYHEASA-N |
Synonym | n-methyl-l-alanine, n-methylalanine, 2s-2-methylamino propanoic acid, h-n-me-ala-oh, l-alanine, n-methyl, alanine, n-methyl, s-n-methylalanine, s-2-methylamino propanoic acid, n-, a-methyl-l-alanine, alanine, n-methyl-, l |
PubChem CID | 5288725 |
ChEBI | CHEBI:17519 |
IUPAC Name | (2S)-2-(methylamino)propanoic acid |
SMILES | CC(C(=O)O)NC |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3913-67-5 | |
103.121 | |
GDFAOVXKHJXLEI-VKHMYHEASA-N | |
5288725 | |
(2S)-2-(methylamino)propanoic acid |
C4H9NO2 | |
MFCD00037241 | |
n-methyl-l-alanine, n-methylalanine, 2s-2-methylamino propanoic acid, h-n-me-ala-oh, l-alanine, n-methyl, alanine, n-methyl, s-n-methylalanine, s-2-methylamino propanoic acid, n-, a-methyl-l-alanine, alanine, n-methyl-, l | |
CHEBI:17519 | |
CC(C(=O)O)NC |
Specifications
3913-67-5 | |
MFCD00037241 | |
n-methyl-l-alanine, n-methylalanine, 2s-2-methylamino propanoic acid, h-n-me-ala-oh, l-alanine, n-methyl, alanine, n-methyl, s-n-methylalanine, s-2-methylamino propanoic acid, n-, a-methyl-l-alanine, alanine, n-methyl-, l | |
CC(C(=O)O)NC | |
103.121 | |
CHEBI:17519 | |
98% |
C4H9NO2 | |
1720922 | |
GDFAOVXKHJXLEI-VKHMYHEASA-N | |
(2S)-2-(methylamino)propanoic acid | |
5288725 | |
103.12 | |
N-Methyl-L-alanine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Methyl-L-alanine, 98%, Thermo Scientific Chemicals