missing translation for 'onlineSavingsMsg'
Learn More

N-n-Butyl-4-nitrobenzamide, 97%, Thermo Scientific™

2060.00 SEK - 6168.00 SEK

Chemical Identifiers

CAS 51207-98-8
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.24
MDL Number MFCD00157824
InChI Key ZPYLRHGQFFDNOH-UHFFFAOYSA-N
Synonym benzamide, n-butyl-4-nitro, n-butyl-4-nitro-benzamide, 4-nitro-n-butylbenzamide, n-n-butyl-4-nitrobenzamide, benzamide, n-butyl-p-nitro, n-butyl 4-nitrophenyl carboxamideShow MoreShow Less
PubChem CID 347929
IUPAC Name N-butyl-4-nitrobenzamide
SMILES CCCCNC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427428
SDS View Documents
Thermo Scientific Alfa Aesar
H60919.MD
250 mg N/A N/A N/A
15417428
SDS View Documents
Thermo Scientific Alfa Aesar
H60919.03
1 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-n-Butyl-4-nitrobenzamide, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.