Learn More
N,N'-Di-n-butylurea, 98%, Thermo Scientific Chemicals
940.00 SEK - 3265.00 SEK
Chemical Identifiers
CAS | 1792-17-2 |
---|---|
Molecular Formula | C9H20N2O |
Molecular Weight (g/mol) | 172.27 |
MDL Number | MFCD00015229 |
InChI Key | AQSQFWLMFCKKMG-UHFFFAOYSA-N |
Synonym | n,n'-dibutylurea, n,n'-di-n-butylurea, urea, n,n'-dibutyl, 1,3-dibutyl urea, unii-j2hug1i10w, j2hug1i10w, n-butyl butylamino carboxamide, nn'-dibutylurea, urea,n'-dibutyl, 1,3-dibutyl-urea |
PubChem CID | 74523 |
IUPAC Name | 1,3-dibutylurea |
SMILES | CCCCNC(=O)NCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11302406
|
Thermo Scientific Alfa Aesar
L06165.06 |
5 g |
940.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312406
|
Thermo Scientific Alfa Aesar
L06165.14 |
25 g |
3265.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N'-Di-n-butylurea is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Di-n-butylurea is used in agrochemical, pharmaceutical and dyestuff field.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Chemical Identifiers
1792-17-2 | |
172.27 | |
AQSQFWLMFCKKMG-UHFFFAOYSA-N | |
74523 | |
CCCCNC(=O)NCCCC |
C9H20N2O | |
MFCD00015229 | |
n,n'-dibutylurea, n,n'-di-n-butylurea, urea, n,n'-dibutyl, 1,3-dibutyl urea, unii-j2hug1i10w, j2hug1i10w, n-butyl butylamino carboxamide, nn'-dibutylurea, urea,n'-dibutyl, 1,3-dibutyl-urea | |
1,3-dibutylurea |
Specifications
1792-17-2 | |
115°C (0.03 mmHg) | |
MFCD00015229 | |
n,n'-dibutylurea, n,n'-di-n-butylurea, urea, n,n'-dibutyl, 1,3-dibutyl urea, unii-j2hug1i10w, j2hug1i10w, n-butyl butylamino carboxamide, nn'-dibutylurea, urea,n'-dibutyl, 1,3-dibutyl-urea | |
CCCCNC(=O)NCCCC | |
172.27 | |
172.27 | |
N,N'-Di-n-butylurea |
69°C to 75°C | |
C9H20N2O | |
1704165 | |
AQSQFWLMFCKKMG-UHFFFAOYSA-N | |
1,3-dibutylurea | |
74523 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 217-258-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.