missing translation for 'onlineSavingsMsg'
Learn More
n-Octadecyltrimethylammonium chloride, 95%, Thermo Scientific Chemicals Product Code.: 15407148

n-Octadecyltrimethylammonium chloride, 95%, Thermo Scientific Chemicals

Product Code. 15407148
2 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 15407148

Brand: Thermo Scientific Alfa Aesar H60665.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Emulsion (main components are high polymeric and cyclic silicones); forms a protective film to coat split ends; conditioning agent for rinses and shampoos; glazing agent for hair.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Emulsion (main components are high polymeric and cyclic silicones); forms a protective film to coat split ends; conditioning agent for rinses and shampoos; glazing agent for hair.

Solubility
Soluble in water 1.759 mg/L @ 25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-03-8
Molecular Formula C21H46ClN
Molecular Weight (g/mol) 348.06
MDL Number MFCD00050188
InChI Key VBIIFPGSPJYLRR-UHFFFAOYSA-M
Synonym n,n,n-trimethyloctadecan-1-aminium chloride, stac, octadecyltrimethylammonium chloride, cation ab, stearyltrimethylammonium chloride, steartrimonium chloride, trimethyloctadecylammonium chloride, 1-octadecanaminium, n,n,n-trimethyl-, chloride, nissan cation ab, aliquat 7Show MoreShow Less
PubChem CID 8155
ChEBI CHEBI:81710
IUPAC Name trimethyl(octadecyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Linear Formula CH3(CH2)17N(Cl)(CH3)3
Quantity 2 g
Beilstein 3917847
Solubility Information Soluble in water 1.759mg/L @ 25°C.
Formula Weight 348.05
Percent Purity 95%
Chemical Name or Material n-Octadecyltrimethylammonium chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ26J023 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog NumberH60665.09,H60665.04
Lot NumberS21J048 Certificate TypeCertificate of Analysis Date27/06/2023 Catalog NumberH60665.09,H60665.04,H60665
Lot Number10178885 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60665.09,H60665.04
Lot NumberW27H024 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60665.09,H60665.04
Lot Number81201872 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60665.09,H60665.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Octadecyltrimethylammonium chloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.