missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl propionate, 99%, Thermo Scientific Chemicals

Product Code. 11489137
Click to view available options
Quantity:
250 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

It has application in automotive refinish, OEM coatings, appliance coatings, printing inks and as polymerization solvent. Its good activity yields efficient resin viscosity reduction that enables the development of lower VOC coatings. Its high electrical resistivity is beneficial in optimizing transfer efficiency in coatings applied via electrostatic spray equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has application in automotive refinish, OEM coatings, appliance coatings, printing inks and as polymerization solvent. Its good activity yields efficient resin viscosity reduction that enables the development of lower VOC coatings. Its high electrical resistivity is beneficial in optimizing transfer efficiency in coatings applied via electrostatic spray equipment.

Solubility
Soluble in water. (5g/L).

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-36-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009373
InChI Key MCSINKKTEDDPNK-UHFFFAOYSA-N
Synonym propyl propionate, n-propyl propionate, propanoic acid, propyl ester, n-propyl propanoate, propionic acid, propyl ester, n-propyl n-propionate, propylester kyseliny propionove, unii-g09trv00gk, propyl propionate natural, propionic acid n-propyl ester
PubChem CID 7803
IUPAC Name propyl propanoate
SMILES CCCOC(=O)CC

Specifications

Melting Point -76°C
Density 0.881
Boiling Point 122°C to 124°C
Flash Point 19°C (67°F)
Linear Formula CH3CH2CO2CH2CH2CH3
Refractive Index 1.393
Quantity 1000 mL
UN Number UN3272
Merck Index 14,7867
Solubility Information Soluble in water. (5g/L).
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material n-Propyl propionate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'