missing translation for 'onlineSavingsMsg'
Learn More
N-t-BOC-Phe-Leu-Phe-Leu-Phe, 97.5%, MP Biomedicals™ Product Code.: 11493911

N-t-BOC-Phe-Leu-Phe-Leu-Phe, 97.5%, MP Biomedicals™

Product Code. 11493911 Shop All MP Biomedical Products
5 mg
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy

Product Code. 11493911

Brand: MP Biomedicals 0215276005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 66556-73-8
Molecular Formula C44H59N5O8
Molecular Weight (g/mol) 785.98
InChI Key NGNZQSPFQJCBJQ-DWCHZDDLSA-N
Synonym boc-phe-d-leu-phe-d-leu-phe-oh, bplplp, butyloxycarbonyl-phenylalanyl-leucyl-phenylalanyl-leucyl-phenylalanine, n-t-boc-phe-leu-phe-leu-phe, n-t-boc-phe-dleu-phe-dleu-phe, butyloxycarbonyl-phe-leu-phe-leu-phe, n-tertiary-butyloxycarbonyl-phenylalanyl-leucyl-phenylalanyl-leucyl-phenylalanine, 2s-2-2r-4-methyl-2-2s-2-2r-4-methyl-2-2s-2-2-methylpropan-2-yl oxycarbonylamino-3-phenyl-propanoyl amino pentanoyl amino-3-phenyl-propanoyl amino pentanoyl amino-3-phenyl-propanoic acid, 2s-2-2r-4-methyl-2-2s-2-2r-4-methyl-2-2s-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoyl amino pentanoyl amino-3-phenylpropanoyl amino pentanoyl amino-3-phenylpropanoic acid, 2s-2-2r-4-methyl-2-2s-2-2r-4-methyl-2-2s-2-2-methylpropan-2-yl oxy-oxomethyl amino-1-oxo-3-phenylpropyl amino-1-oxopentyl amino-1-oxo-3-phenylpropyl amino-1-oxopentyl amino-3-phenylpropanoic acidShow MoreShow Less
PubChem CID 125628

Specifications

Quantity 5 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-t-BOC-Phe-Leu-Phe-Leu-Phe, 97.5%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.