missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N1-(4-acetyl-3-hydroxyphenyl)acetamide, Thermo Scientific™
2075.00 SEK - 6655.00 SEK
Chemical Identifiers
CAS | 40547-58-8 |
---|---|
Molecular Formula | C10H11NO3 |
Molecular Weight (g/mol) | 193.202 |
InChI Key | ZSSHNMZQDWSUJJ-UHFFFAOYSA-N |
Synonym | n-4-acetyl-3-hydroxyphenyl acetamide, n1-4-acetyl-3-hydroxyphenyl acetamide, 4'-acetamido-2'-hydroxyacetophenone, acetamide, n-4-acetyl-3-hydroxyphenyl, 4-acetamido-2-hydroxyacetophenone, n-4-acetyl-3-hydroxy-phenyl acetamide, n-4-acetyl-3-hydroxy-phenyl-acetamide |
PubChem CID | 2775185 |
IUPAC Name | N-(4-acetyl-3-hydroxyphenyl)acetamide |
SMILES | CC(=O)C1=C(C=C(C=C1)NC(=O)C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10118122
|
Thermo Scientific Maybridge
BTB10058DA |
1 g |
2075.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10096984
|
Thermo Scientific Maybridge
BTB10058DE |
5 g |
6655.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
40547-58-8 | |
193.202 | |
n-4-acetyl-3-hydroxyphenyl acetamide, n1-4-acetyl-3-hydroxyphenyl acetamide, 4'-acetamido-2'-hydroxyacetophenone, acetamide, n-4-acetyl-3-hydroxyphenyl, 4-acetamido-2-hydroxyacetophenone, n-4-acetyl-3-hydroxy-phenyl acetamide, n-4-acetyl-3-hydroxy-phenyl-acetamide | |
N-(4-acetyl-3-hydroxyphenyl)acetamide |
C10H11NO3 | |
ZSSHNMZQDWSUJJ-UHFFFAOYSA-N | |
2775185 | |
CC(=O)C1=C(C=C(C=C1)NC(=O)C)O |
Specifications
40547-58-8 | |
n-4-acetyl-3-hydroxyphenyl acetamide, n1-4-acetyl-3-hydroxyphenyl acetamide, 4'-acetamido-2'-hydroxyacetophenone, acetamide, n-4-acetyl-3-hydroxyphenyl, 4-acetamido-2-hydroxyacetophenone, n-4-acetyl-3-hydroxy-phenyl acetamide, n-4-acetyl-3-hydroxy-phenyl-acetamide | |
CC(=O)C1=C(C=C(C=C1)NC(=O)C)O | |
193.202 | |
193.2 | |
Technical |
C10H11NO3 | |
ZSSHNMZQDWSUJJ-UHFFFAOYSA-N | |
N-(4-acetyl-3-hydroxyphenyl)acetamide | |
2775185 | |
Tech. | |
N1-(4-Acetyl-3-hydroxyphenyl)acetamide |