missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Scientific 2.5mL 15 x 45mm Crimp/Snap-Ring LoVials™

Feature a 13mm crimp/snap-ring finish, for use with aluminum seals or Kim-Snap™ closures.

Brand:  Thermo Scientific™ C4015-12

Product Code. 12430178

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ National 2.5mL 15 x 45mm Crimp/Snap-Ring LoVials feature a 13mm crimp/snap-ring finish, for use with aluminum seals or Kim-Snap™ closures.

  • Fits Waters'™ 4mL WISP™ and other 15 × 45mm autosampler trays
  • Vials feature a 13mm crimp/snap-ring finish—use with Aluminum Seals or Kim-Snap™ Closures
  • Superior quality 33 expansion borosilicate clear glass Type 1, Class A Uniformly flat bottom for security with inserts
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene
<300 μL
13 mm
2.5 mL
2.5 mL
Aluminum Seals or Kim-Snap™ Closures
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers