Learn More
Abeba™ Nature 8045 Shoes
Leather covered cork footbed is ESD dissipative.
Brand:  Abeba™ 8045/45
 View more versions of this product
Product Code. 15642998
  • 974.00 SEK / 1 pair
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
45
Rubber
Black
Leather
Slip-resistant
8045