Learn More
Abeba™ Nature 8045 Shoes
Leather covered cork footbed is ESD dissipative.
Brand:  Abeba™ 8045/47
 View more versions of this product
Product Code. 15662998
  • 974.00 SEK / 1 pair
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
47
Rubber
Black
Leather
Slip-resistant
8045