missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCF4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1905.00 SEK - 4225.00 SEK
Specifications
Antigen | NCF4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18323633
|
Bio-Techne
NBP3-16942-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18373183
|
Bio-Techne
NBP3-16942-100UL |
100 μg |
4225.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NCF4 Polyclonal antibody specifically detects NCF4 in Human samples. It is validated for Western BlotSpecifications
NCF4 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
MGC3810, NCF, NCF-4, neutrophil cytosol factor 4, neutrophil cytosolic factor 4 (40kD), neutrophil cytosolic factor 4, 40kDa, Neutrophil NADPH oxidase factor 4, p40phox, p40-phox, SH3 and PX domain-containing protein 4, SH3PXD4P40PHOX | |
A synthetic phosphorylated peptide around T154 of human NCF4 (NP_000622.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 50% glycerol, pH7.3 | |
4689 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title