missing translation for 'onlineSavingsMsg'
Learn More

NDUFB8 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

3675.00 SEK - 5775.00 SEK

Specifications

Antigen NDUFB8
Dilution Western Blot 0.04-0.4 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18323674
View Documents
Bio-Techne
NBP3-17898-25UL
25 μg
3675.00 SEK
25µL
Please to purchase this item. Need a web account? Register with us today!
18378503
View Documents
Bio-Techne
NBP3-17898-100UL
100 μg
5775.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NDUFB8 Polyclonal antibody specifically detects NDUFB8 in Human samples. It is validated for Western Blot
Specifications

Specifications

NDUFB8
Western Blot
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS, pH 7.2, 40% glycerol
4714
IgG
Affinity purified
Western Blot 0.04-0.4 ug/ml
Polyclonal
Purified
RUO
Human
CI-ASHImitochondrial, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8 (19kD, ASHI), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8, 19kDa, NADH:ubiquinone oxidoreductase ASHI subunit, NADH-ubiquinone oxidoreductase ASHI subunit
This antibody was developed against Recombinant Protein corresponding to amino acids: YPVYQPVGPKQYPYNNLYLERGGDPSKEPERVVHYEI
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.