Learn More
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UF110plus
Nearly gastight interior with possibility for inlet/outlet through two tubes with ball valves.
Brand:  Memmert™ UF110plusK3
Product Code. 17548985
  • 10820.00 SEK / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Gas inlet/outlet through 2 tubes with ball valves
  • Technical clarification required for combination with other options
Specifications
Specifications
UF110plus Oven
Interior
Gas Inlet/Outlet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UF110plus > For Use With: UF110plus