Learn More
Eppendorf™ Test Tube Racks for Model C76 Water Bath Shaker
Test Tube Racks have an adjustable angle and tube height support. Made from stainless-steel with inserts to help retain tubes and prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Brand: Eppendorf™ AG7-TT25
This item is not returnable.
View return policy
Description
Require subplatform (14-284-46); order separately.
Specifications
C76 water bath shaker | |
18.0 25 mm Tubes | |
Insert | |
Stainless Steel |