missing translation for 'onlineSavingsMsg'
Learn More

NF45 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen NF45
Concentration 0.13 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883494
View Documents Promotion Details
Proteintech
14714-1-AP-20UL
20 μL
1656.73 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16873494
View Documents Promotion Details
Proteintech
14714-1-AP-150UL
150 μL
4721.67 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nuclear factor of activated T-cells is a transcription factor required for T-cell expression of the interleukin 2 gene. NFAT binds to a sequence in the interleukin 2 gene enhancer known as the antigen receptor response element 2. In addition, NFAT can bind RNA and is an essential component for encapsidation and protein priming of hepatitis B viral polymerase. NFAT is a heterodimer of 45 kDa and 90 kDa proteins, the smaller of which is the product of this gene. The encoded protein binds strongly to the 90 kDa protein and stimulates its ability to enhance gene expression.
Specifications

Specifications

NF45
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Human
Q12905, Q9CXY6
3608, 67781
NF45 Fusion Protein Ag6436
Primary
-20°C
ILF2
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ILF2, NF45, PRO3063
ILF2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications