Learn More
Cole-Parmer™ Nickel-plated Brass Luer Fittings
Brand: Cole-Parmer™ 41507-70
Additional Details : Weight : 0.02000kg
Specifications
Luer Fitting | |
Nickle-plated Brass | |
Harsh chemical environments | |
10-32 UNF Thread |
Autoclavable | |
Male Luer Lock | |
1 Pack |